EMAIL THIS PAGE TO A FRIEND
Frontiers in microbiology
Characterization of Bile Salt Hydrolase from
PMID 28620369
Abstract
Probiotic bacteria are beneficial to the health of poultry animals, thus are used as alternative candidates for antibiotics used as growth promoters (AGPs). However, they also reduce the body weight gain due to innate bile salt hydrolase (BSH) activity. Hence, the addition of a suitable BSH inhibitor along with the probiotic feed can decrease the BSH activity. In this study, a BSH gene (981 bp) encoding 326-amino acids was identified from the genome of
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B3809
Benzenesulfonyl hydrazide, 98%
C6H8N2O2S