EMAIL THIS PAGE TO A FRIEND
Veterinary microbiology
Brucella lipopolysaccharide reinforced Salmonella delivering Brucella immunogens protects mice against virulent challenge.
PMID 28622869
Abstract
Intracellular pathogen Salmonella exhibits natural infection broadly analogous to Brucella, this phenomenon makes Salmonella a pragmatic choice for an anti-Brucella vaccine delivery platform. In this study we developed and formulated a combination of four attenuated Salmonella Typhimurium live vector strains delivering heterologous Brucella antigens (rBs), namely lumazine synthase, proline racemase subunit A, lipoprotein outer membrane protein-19, and Cu-Zn superoxide dismutase. With an aim to develop a cross-protecting vaccine, Brucella pan-species conserved rBs were selected. The present study compared the efficacy of smooth and rough variants of Salmonella delivery vector and also evaluated the inclusion of purified Brucella lipopolysaccharide (LPS) in the formulation. Immunization of SPF-BALB/c mice with the vaccine combinations significantly (P≤0.05) reduced splenic wild-type Brucella abortus 544 colonization as compared to non-immunized mice as well as Salmonella only immunized mice. Increased induction of Brucella specific-IgG, sIgA production, and antigen-specific splenocyte proliferative responses were observed in the mice immunized with the formulations as compared to naïve or vector only immunized mice. Modulatory effects of rB and LPS on production of interleukin (IL)-4, IL-12, and interferon-γ were detected in splenocytes of mice immunized with the formulation. Rough Salmonella variant in combination with LPS could further enhance the efficacy of the delivery when applied intraperitoneally. Taken together, it is compelling that Brucella LPS-augmented Salmonella vector delivering immunogenic Brucella proteins may be more suitable than the current non-ideal live Brucella abortus vaccine. The vaccine system also provides a basis for the development of cross-protecting vaccine capable of preventing multispecies brucellosis.