EMAIL THIS PAGE TO A FRIEND
Drug and alcohol dependence
HIV Tat excites D1 receptor-like expressing neurons from rat nucleus accumbens.
PMID 28623807
Abstract
HIV-1 infection and drug abuse are frequently co-morbid and their association greatly increases the severity of HIV-1-induced neuropathology. While nucleus accumbens (NAcc) function is severely perturbed by drugs of abuse, little is known about how HIV-1 infection affects NAcc. We used calcium and voltage imaging to investigate the effect of HIV-1 trans-activator of transcription (Tat) on rat NAcc. Based on previous neuronal studies, we hypothesized that Tat modulates intracellular Ca We provide evidence that Tat triggers a Ca We describe for the first time a HIV-1 Tat-triggered Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8189
Bis(1,3-dibutylbarbituric acid) trimethine oxonol, ≥95% (HPLC)
C27H40N4O6
S1148
SBFI-AM
C56H58N2O23