EMAIL THIS PAGE TO A FRIEND
Journal of dairy science
Effect of endoplasmic reticulum stress on metabolic and stress signaling and kidney-specific functions in Madin-Darby bovine kidney cells.
PMID 28624282
Abstract
Recent studies demonstrated induction of endoplasmic reticulum (ER) stress in tissues of cows after parturition, but knowledge about the effect of ER stress on important cellular processes, such as critical signaling and metabolic pathways, in cattle is scarce. Thus, the present study aimed to investigate the effect of ER stress induction on nuclear factor-κB (NF-κB), nuclear factor E2-related factor 2 (Nrf2), and sterol regulatory element-binding protein (SREBF1) pathway in Madin-Darby bovine kidney (MDBK) cells, a widely used in vitro model in ruminant research. To consider the kidney origin of MDBK cells, the effect on renal distal tubular cell-specific functions, such as transport processes and regulation of 1,25(OH)