EMAIL THIS PAGE TO A FRIEND
Free radical biology & medicine
Silymarin protects against renal injury through normalization of lipid metabolism and mitochondrial biogenesis in high fat-fed mice.
PMID 28625483
Abstract
Obesity is associated with an increased risk of chronic kidney diseases and the conventional treatment with renin-angiotensin-aldosterone system (RAAS) inhibitors is not enough to prevent renal injury and prolong the progression of disease. Recently, silymarin has shown protective effects on renal tissue injury, but the underlying mechanisms remain elusive. The goal of this study was to investigate the potential capacity of silymarin to prevent renal injury during obesity induced by high fat diet (HFD) in mice. In vivo, male C57BL/6 mice received HFD (60% of total calories) for 12 weeks, randomized and treated orally with vehicle saline or silymarin (30mg/kg body weight/d) for 4 weeks. In vitro, human proximal tubular epithelial cells (HK2) were exposed to 300μM palmitic acid (PA) for 36h followed by silymarin administration at different concentrations. The administration of silymarin significantly ameliorated HFD induced glucose metabolic disorders, oxidative stress and pathological alterations in the kidney. Silymarin significantly mitigated renal lipid accumulation, fatty acid β-oxidation and mitochondrial biogenesis in HFD mice and PA treated HK2 cells. Furthermore, silymarin partly restored mitochondrial membrane potential of HK2 cells after PA exposure. In conclusion, silymarin can improve oxidative stress and preserve mitochondrial dysfunction in the kidney, potentially via preventing accumulation of renal lipids and fatty acid β-oxidation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S0292 Silymarin, flavonolignans