EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences
Phospholipid-drug conjugates as a novel oral drug targeting approach for the treatment of inflammatory bowel disease.
PMID 28627471
Abstract
The enzyme phospholipase A
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
217700
DL-2-Aminocaprylic acid, 99%
C8H17NO2