EMAIL THIS PAGE TO A FRIEND
Environmental science & technology
Secondary Organic Aerosol (SOA) from Nitrate Radical Oxidation of Monoterpenes: Effects of Temperature, Dilution, and Humidity on Aerosol Formation, Mixing, and Evaporation.
PMID 28628304
Abstract
Nitrate radical (NO