EMAIL THIS PAGE TO A FRIEND
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
MiR-495 Promotes Senescence of Mesenchymal Stem Cells by Targeting Bmi-1.
PMID 28628915
Abstract
Mesenchymal stem cells (MSCs) play an important role in regulating angiogenesis and immune balance. Abnormal proliferation and function of MSCs were reported at maternal fetal interface in patients with pre-eclampsia (PE). Micro-RNA-495 was known to be upregulated in the MSCs derived from patients with PE. However, it is not clear whether the up-regulated miR-495 is related to the pathogenesis of PE. We analyzed the expression of miR-495 in MSCs and umbilical cords derived from healthy pregnancies (NC) and PE, then we upregulated or downregulated the expression of miR-495 in MSCs derived from NC and tested the proliferation, apoptosis, migration, invasion, tube formation and senescence. In the current study, we found that the expression of miR-495 was significantly increased in both umbilical cord tissues and MSCs in patients with severe PE. Overexpressing miR-495 arrested cell cycle in S phase and promoted cell apoptosis. The supernatants from miR-495-overexpressed-MSCs inhibited the migration of MSCs and HTR-8/SVneo, invasion of HTR-8/SVneo and tube formation of HUVEC, while si-miR-495 had the opposite effects. Furthermore, we analyzed the senescence related β-galactosidase activity and CD146 and found that miR-495 induced the senescence of MSCs. Molecular mechanism studies confirmed that Bmi-1 mediated these effects of miR-495 on MSCs. Taken together, our data demonstrated that miR-495 induced senescence of MSCs may be involved in the pathogenesis of PE.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295272
1,1-Difluoroethylene, ≥99%
C2H2F2