EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Biodiesel synthesis from Calophyllum inophyllum oil with different supercritical fluids.
PMID 28628981
Abstract
Biodiesel or fatty acid methyl esters (FAMEs) is primarily synthesized using edible vegetable oils and methanol with a catalyst. However, in the present study, FAMEs were synthesized from a non-edible oil (Calophyllum inophyllum also called as sura honne, Punnagam, Alexandrian Laurel) in different supercritical fluids: methanol (MeOH), methyl tert-butyl ether (MTBE), methyl acetate (MeOAc) and dimethyl carbonate (DMC) non-catalytically. Reactions were performed from 523K to 673K at 30MPa with a molar ratio of 40:1 with times varying from 3min to 3h. Conversions higher than 80% were obtained within 30min for oil reaction with MeOH and DMC at 623K and conversions of 60% and 70% were obtained at 673K with MeOAc and MTBE, respectively. Pseudo first order kinetics was used to obtain the rate constants and the activation energies followed the order: E
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W220604
Butyl laurate, ≥99%
C16H32O2