EMAIL THIS PAGE TO A FRIEND
Materials science & engineering. C, Materials for biological applications
A colon targeted drug delivery system based on alginate modificated graphene oxide for colorectal liver metastasis.
PMID 28629006
Abstract
A major problem associated with colon cancer is liver metastasis. A colon-targeted drug delivery system is one way to address this problem after the resection of colorectal cancer. However, traditional drug delivery systems face many challenges, such as an inability to control the release rate, inaccurate targeting, susceptibility to the microenvironment and poor stability. Here, we report the development of a graphene oxide (GO)-based, sodium alginate (ALG) functionalized colon-targeting drug delivery system, that is loaded with 5-fluorouracil (5-FU) as the anti-cancer drug (denoted as GO-ALG/5-FU). Our results demonstrate that the as-prepared drug delivery system possesses a much lower toxicity and better colon-targeting controlled-release behaviours. We show that GO-ALG/5-FU significantly inhibited tumour growth and liver metastasis and prolonged the survival time of mice. We anticipate that our assay will help improve basic research of colon-targeted drug delivery systems and provide a new way to treat colon cancer liver metastasis.