EMAIL THIS PAGE TO A FRIEND
Aging
The association between telomere length and mortality in Bangladesh.
PMID 28630379
Abstract
Telomeres are tandem repeat sequences at the end of chromosomes that bind proteins to protect chromosome ends. Telomeres shorten with age, and shorter leukocyte telomere length (TL) has been associated with overall mortality in numerous studies. However, this association has not been tested in populations outside of Europe and the U.S. We assessed the association between TL and subsequent mortality using data on 744 mortality cases and 761 age-/sex-matched controls sampled from >27,000 participants from three longitudinal Bangladeshi cohorts: Health Effects of Arsenic Longitudinal Study (HEALS), HEALS Expansion (HEALS-E), and Bangladesh Vitamin E and Selenium Trial (BEST). We used conditional logistic regression to estimate odds ratios (ORs) for the association between a standardized TL variable and overall mortality, as well as mortality from chronic diseases, respiratory diseases, circulatory diseases, and cancer. In HEALS and BEST, we observed an association between shorter TL and increased overall mortality (P=0.03 and P=0.03), mortality from chronic disease (P=0.01 and P=0.03) and mortality from circulatory disease (P=0.03 and P=0.04). Results from pooled analyses of all cohorts were consistent with HEALS and BEST. This is the first study demonstrating an association between short TL and increased mortality in a population of non-European ancestry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
364681
2-(Trimethylsilyl)ethanethiol, 95%
C5H14SSi