EMAIL THIS PAGE TO A FRIEND
Chemistry, an Asian journal
Tuning the Surface Polarity of Microporous Organic Polymers for CO
PMID 28631882
Abstract
CO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T82007
1,3,5-Triphenylbenzene, 97%
C24H18