EMAIL THIS PAGE TO A FRIEND
Ultrasonics sonochemistry
Effects of pulsed ultrasound on rheological and structural properties of chicken myofibrillar protein.
PMID 28633822
Abstract
The effects of pulsed ultrasound (PUS) (power: 240w) with varying time (0, 3, 6, 9, 12 and 15min) on rheological and structural properties of chicken myofibrillar protein (CMP) were examined. PUS treatment significantly caused a decrease in the viscosity coefficients (k) but an increase in the flow index (n) value of CMP solutions within short time (0-6min), while had no significant effect for longer time (9-15min). Besides, at 6min, the solubility and microstructure of CMP samples were optimum. The primary structure of CMP was not altered by PUS treatment. However, Raman spectroscopy revealed a decrease in the α-helix and β-sheets proportion and an increase in the β-turn of CMP following PUS treatment. Random coil reached a maximum at 6min. The changes in tertiary and quaternary structure of CMP by PUS treatment also occurred. As PUS time extended, S
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
186341
1-Naphthalenesulfonic acid, >50%
C10H8O3S