EMAIL THIS PAGE TO A FRIEND
Nanomedicine (London, England)
Multimodal imaging Gd-nanoparticles functionalized with Pittsburgh compound B or a nanobody for amyloid plaques targeting.
PMID 28635419
Abstract
Gadolinium-based nanoparticles were functionalized with either the Pittsburgh compound B or a nanobody (B10AP) in order to create multimodal tools for an early diagnosis of amyloidoses. The ability of the functionalized nanoparticles to target amyloid fibrils made of β-amyloid peptide, amylin or Val30Met-mutated transthyretin formed in vitro or from pathological tissues was investigated by a range of spectroscopic and biophysics techniques including fluorescence microscopy. Nanoparticles functionalized by both probes efficiently interacted with the three types of amyloid fibrils, with K Such functionalized nanoparticles could represent promising flexible and multimodal imaging tools for the early diagnostic of amyloid diseases, in other words, Alzheimer's disease, Type 2 diabetes mellitus and the familial amyloidotic polyneuropathy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T1892 Thioflavine S, practical grade