Bioconjugate chemistry
A2B-Miktoarm Glycopolymer Fibers and Their Interactions with Tenocytes.
PMID 28636335
Electrospun biodegradable membranes have attracted great attention for a range of tissue engineering applications. Among them, poly(ε-caprolactone) (PCL) is one of the most widely used polymers, owing to its well-controlled biocompatibility and biodegradability. However, PCL also has a number of limitations, such as its hydrophobic nature and the lack of functional groups on its side chain, limiting its ability to interact with cells. Herein, we have designed and prepared a series of well-defined A2B-miktoarm copolymers with PCL and glycopolymer segments to address these limitations. Moreover, copolymers were electrospun to make membranes, which were studied in vitro to investigate cell affinity, toxicity, activity, and adhesion with these materials. The results indicate that incorporating glucose moieties into miktoarm polymers has improved the biocompatibility of the PCL while increasing the cellular interaction with the membrane material.