EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Cell physiology
Oxidative stress destabilizes protein arginine methyltransferase 4 via glycogen synthase kinase 3β to impede lung epithelial cell migration.
PMID 28637674
Abstract
Oxidative stress impacts normal cellular function leading to the pathogenesis of various diseases including pulmonary illnesses. Protein arginine methyltransferase 4 (PRMT4) is critical for normal lung alveolar epithelial cell development; however, the regulation of PRMT4 within such pulmonary diseases has yet to be elucidated. Using biochemical approaches, we uncovered that peroxide (H
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1053
O-Phospho-L-threonine
C4H10NO6P