EMAIL THIS PAGE TO A FRIEND
Plant physiology
A Light Harvesting Complex-Like Protein in Maintenance of Photosynthetic Components in
PMID 28637830
Abstract
Using a genetic approach, we have identified and characterized a novel protein, named Msf1 (Maintenance factor for photosystem I), that is required for the maintenance of specific components of the photosynthetic apparatus in the green alga