EMAIL THIS PAGE TO A FRIEND
Nature communications
MicroRNA-34a dependent regulation of AXL controls the activation of dendritic cells in inflammatory arthritis.
PMID 28639625
Abstract
Current treatments for rheumatoid arthritis (RA) do not reverse underlying aberrant immune function. A genetic predisposition to RA, such as HLA-DR4 positivity, indicates that dendritic cells (DC) are of crucial importance to pathogenesis by activating auto-reactive lymphocytes. Here we show that microRNA-34a provides homoeostatic control of CD1c