EMAIL THIS PAGE TO A FRIEND
Food & function
Octyl gallate, a food additive with potential beneficial properties to treat Helicobacter pylori infection.
PMID 28640317
Abstract
Helicobacter pylori infection is marked by intense production of reactive oxygen species (ROS) through the activation of neutrophils that are constantly attracted to the infected gastric mucosa. Here, gallic acid and its alkyl esters were evaluated as compounds able to act as antimicrobial agents and inhibitors of ROS released by H. pylori-activated neutrophils simultaneously. We found that the higher hydrophobicity caused by esterification of gallic acid led to a significant increase in its ability as a cytotoxic agent against H. pylori, a scavenger of ROS and an inhibitor of NADPH oxidase in neutrophils. Octyl gallate, a widely used food additive, showed the highest antimicrobial activity against H. pylori, with a minimum inhibitory concentration (MIC) value of 125 μg mL
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
274194
Methyl 3,4,5-trihydroxybenzoate, 98%
C8H8O5