EMAIL THIS PAGE TO A FRIEND
International journal of food microbiology
Impact of Saccharomyces cerevisiae and Torulaspora delbrueckii starter cultures on cocoa beans fermentation.
PMID 28641144
Abstract
Aim of this work was to study the impact of mixed cultures of Saccharomyces cerevisiae and Torulaspora delbrueckii and T. delbrueckii monoculture on the fermentation process conducted on two different cocoa hybrids, PS1319 and SJ02, in Bahia, Brazil. This was performed throughout studying physico-chemical changes during the fermentation process and analyzing volatile compounds and sensory analysis of chocolates. (GTG)