EMAIL THIS PAGE TO A FRIEND
Cancer immunology research
Modulation of Endoplasmic Reticulum Stress Controls CD4
PMID 28642246
Abstract
The endoplasmic reticulum (ER) is an energy-sensing organelle with intimate ties to programming cell activation and metabolic fate. T-cell receptor (TCR) activation represents a form of acute cell stress and induces mobilization of ER Ca