EMAIL THIS PAGE TO A FRIEND
Scientific reports
Comparing DNA enrichment of proliferating cells following administration of different stable isotopes of heavy water.
PMID 28642474
Abstract
Deuterated water (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7400
2′-Deoxyadenosine monohydrate, ≥99%
C10H13N5O3 · H2O