EMAIL THIS PAGE TO A FRIEND
Nature chemistry
Redox-controlled potassium intercalation into two polyaromatic hydrocarbon solids.
PMID 28644481
Abstract
Alkali metal intercalation into polyaromatic hydrocarbons (PAHs) has been studied intensely after reports of superconductivity in a number of potassium- and rubidium-intercalated materials. There are, however, no reported crystal structures to inform our understanding of the chemistry and physics because of the complex reactivity of PAHs with strong reducing agents at high temperature. Here we present the synthesis of crystalline K
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
236705
Pentane, anhydrous, ≥99%
C5H12