EMAIL THIS PAGE TO A FRIEND
Journal of plant physiology
Overexpression of Populus×canescens isoprene synthase gene in Camelina sativa leads to alterations in its growth and metabolism.
PMID 28644970
Abstract
Isoprene (2-methyl-1,3-butadiene) is a hemiterpene molecule. It has been estimated that the plant kingdom emits 500-750 million tons of isoprene in the environment, half of which results from tropical broadleaf trees and the remainder from shrubs. Camelina (Camelina sativa (L.) Crantz) is an emerging bioenergy plant for biodiesel. In this study, we characterized isoprene formation following a diurnal/nocturnal cycle in wild-type Camelina plants. To understand the potential effects of isoprene emission on this herbaceous plant, a gray poplar Populus×canescens isoprene synthase gene (PcISPS) was overexpressed in Camelina. Transgenic plants showed increased isoprene production, and the emissions were characterized by a diurnal/nocturnal cycle. Measurements of the expression of six genes of the plastidial 2-C-methyl-d-erythriol-4-phosphate (MEP) pathway revealed that the expression patterns of three key genes were associated with isoprene formation dynamics in the three genotypic plants. Conversely, dissimilar gene expression levels existed in different genotypes, indicating that dynamics and variations occurred among plants. Moreover, transgenic plants grew shorter and developed smaller leaves than the wild-type and empty vector control transgenic plants. Photosynthetic analysis showed that the CO