EMAIL THIS PAGE TO A FRIEND
Molecular pharmacology
A Functional Na
PMID 28645932
Abstract
The Na
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
746959
Ethenesulfonyl fluoride, 95%
C2H3FO2S