EMAIL THIS PAGE TO A FRIEND
Drug and alcohol dependence
Synthetic cathinones and stereochemistry: S enantiomer of mephedrone reduces anxiety- and depressant-like effects in cocaine- or MDPV-abstinent rats.
PMID 28646714
Abstract
The neuropharmacological profile of the synthetic cathinone mephedrone (MEPH) is influenced by stereochemistry. Both MEPH enantiomers are monoamine transporter substrates, but R-MEPH is primarily responsible for rewarding effects of MEPH as it produces greater locomotor activation and intracranial self-stimulation than S-MEPH. S-MEPH is a 50-fold more potent 5-HT releaser than R-MEPH and does not place preference in rats. MEPH is also structurally similar to the cathinone derivative bupropion, an antidepressant and smoking cessation medication, suggesting MEPH has therapeutic and addictive properties. We tested the hypothesis that S-MEPH reduces anxiety- and depression-like behaviors in rats withdrawn from chronic cocaine or methylenedioxypyrovalerone (MDPV) using the elevated plus maze (EPM) and forced swim test (FST), respectively. Rats were tested 48-h after a binge-like paradigm (3×/day for 10days in 1-h intervals) of cocaine (10mg/kg), MDPV (1mg/kg) or saline. In vitro studies assessed the receptor binding and activity of S-MEPH. Rats withdrawn from chronic cocaine or MDPV displayed an increase in anxiety- and depression-like behaviors that was antagonized by treatment with S-MEPH (10, 30mg/kg). S-MEPH displayed affinity, but not agonist activity, for 5-HT S-MEPH attenuated withdrawal behaviors following chronic cocaine or MDPV, perhaps through 5-HT release and/or 5-HT