Journal of neuroinflammation
Influence of type I IFN signaling on anti-MOG antibody-mediated demyelination.
PMID 28646890
Antibodies with specificity for myelin oligodendrocyte glycoprotein (MOG) are implicated in multiple sclerosis and related diseases. The pathogenic importance of anti-MOG antibody in primary demyelinating pathology remains poorly characterized. The objective of this study is to investigate whether administration of anti-MOG antibody would be sufficient for demyelination and to determine if type I interferon (IFN) signaling plays a similar role in anti-MOG antibody-mediated pathology, as has been shown for neuromyelitis optica-like pathology. Purified IgG2a monoclonal anti-MOG antibody and mouse complement were stereotactically injected into the corpus callosum of wild-type and type I IFN receptor deficient mice (IFNAR1-KO) with and without pre-established experimental autoimmune encephalomyelitis (EAE). Anti-MOG induced complement-dependent demyelination in the corpus callosum of wild-type mice and did not occur in mice that received control IgG2a. Deposition of activated complement coincided with demyelination, and this was significantly reduced in IFNAR1-KO mice. Co-injection of anti-MOG and complement at onset of symptoms of EAE induced similar levels of callosal demyelination in wild-type and IFNAR1-KO mice. Anti-MOG antibody and complement was sufficient to induce callosal demyelination, and pathology was dependent on type I IFN. Induction of EAE in IFNAR1-KO mice overcame the dependence on type I IFN for anti-MOG and complement-mediated demyelination.