EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Practical, Broadly Applicable, α-Selective, Z-Selective, Diastereoselective, and Enantioselective Addition of Allylboron Compounds to Mono-, Di-, Tri-, and Polyfluoroalkyl Ketones.
PMID 28648084
Abstract
A practical method for enantioselective synthesis of fluoroalkyl-substituted Z-homoallylic tertiary alcohols has been developed. Reactions may be performed with ketones containing a polylfluoro-, trifluoro-, difluoro-, and monofluoroalkyl group along with an aryl, a heteroaryl, an alkenyl, an alkynyl, or an alkyl substituent. Readily accessible unsaturated organoboron compounds serve as reagents. Transformations were performed with 0.5-2.5 mol % of a boron-based catalyst, generated in situ from a readily accessible valine-derived aminophenol and a Z- or an E-γ-substituted boronic acid pinacol ester. With a Z organoboron reagent, additions to trifluoromethyl and polyfluoroalkyl ketones proceeded in 80-98% yield, 97:3 to >98:2 α:γ selectivity, >95:5 Z:E selectivity, and 81:19 to >99:1 enantiomeric ratio. In notable contrast to reactions with unsubstituted allylboronic acid pinacol ester, additions to ketones with a mono- or a difluoromethyl group were highly enantioselective as well. Transformations were similarly efficient and α- and Z-selective when an E-allylboronate compound was used, but enantioselectivities were lower. In certain cases, the opposite enantiomer was favored (up to 4:96 er). With a racemic allylboronate reagent that contains an allylic stereogenic center, additions were exceptionally α-selective, affording products expected from γ-addition of a crotylboron compound, in up to 97% yield, 88:12 diastereomeric ratio, and 94:6 enantiomeric ratio. Utility is highlighted by gram-scale preparation of representative products through transformations that were performed without exclusion of air or moisture and through applications in stereoselective olefin metathesis where Z-alkene substrates are required. Mechanistic investigations aided by computational (DFT) studies and offer insight into different selectivity profiles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
907928
(S)-2-((2-Hydroxy-3-(triphenylsilyl)benzyl)amino)-N,N,3-trimethylbutanamide, ≥95%
C32H36N2O2Si