EMAIL THIS PAGE TO A FRIEND
Food microbiology
Light based technologies for microbial inactivation of liquids, bead surfaces and powdered infant formula.
PMID 28648293
Abstract
This study evaluates the potential of continuous wave Ultraviolet C light (UV-C) and broad-spectrum intense pulsed light (in this study referred to as High Intensity Light Pulses, HILP) for the inactivation of pathogens of public concern in powdered infant formula (PIF) producers. To achieve this goal a sequential set of experiments were performed, firstly in clear liquid media, secondly on the surface of spherical beads under agitation and, finally in PIF. L. innocua was the most sensitive microorganism to both technologies under all conditions studied with reductions exceeding 4 log
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14703 Cronobacter spp. ChromoSelect Agar, modified, for microbiology