EMAIL THIS PAGE TO A FRIEND
Environmental pollution (Barking, Essex : 1987)
Quantification and speciation of volatile fatty acids in the aqueous phase.
PMID 28649044
Abstract
This study lays great emphasis on establishing a reliable analytical platform to quantify and specify volatile fatty acids (VFAs) in the aqueous phase by derivatizing VFAs into their corresponding alkyl esters via thermally-induced rapid esterification (only 10 s reaction time). To this end, reaction conditions for the thermally-induced rapid esterification are optimized. A volumetric ratio of 0.5 at 400 °C for VFA/methanol is identified as the optimal reaction conditions to give ∼90% volatile fatty acid methyl ester (VFAME) yield. To maintain a high yield of VFAMEs, this study suggests that dilution of the sample to an optimum concentration (∼500 ppm for each VFA) is required. Derivatization of VFAs into VFAMEs via the thermally-induced rapid esterification is more reliable to quantify and specify VFAs in the aqueous phase than conventional colorimetric method.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W310212
Isovaleric acid, natural, ≥98%, FG
C5H10O2
59900
Methyl isovalerate, ≥98.0% (GC)
C6H12O2