EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Ligand- and Solvent-Tuned Chemoselective Carbonylation of Bromoaryl Triflates.
PMID 28650074
Abstract
The palladium-catalyzed chemoselective carbonylation of bromoaryl triflates is reported. The selective C-Br bond versus C-OTf (OTf=triflate) bond functionalization can be remarkably tuned by the combination of the ligand [4,5-bis(diphenylphosphino)-9,9-dimethylxanthene (Xantphos) vs. 1,1'-bis(diphenylphosphino)ferrocene (DPPF)] and the solvent (toluene vs. DMSO). The respective ligand and solvent effects are rationalized by DFT calculations. In contrast, the monodentate ligands BuPAd
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
138576
1-Adamantylamine, 97%
C10H17N
B73201
1-Bromo-2-naphthol, 97%
C10H7BrO
B73406
6-Bromo-2-naphthol, 97%
C10H7BrO