EMAIL THIS PAGE TO A FRIEND
Environmental Pollution (Barking, Essex : 1987)
Genetic and epigenetic alterations in normal and sensitive COPD-diseased human bronchial epithelial cells repeatedly exposed to air pollution-derived PM2.5.
PMID 28651088