EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Effect of salt type and concentration on the growth and lipid content of Chlorella vulgaris in synthetic saline wastewater for biofuel production.
PMID 28651134
Abstract
Microalgae can offer several benefits for wastewater treatment with their ability to produce large amounts of lipids for biofuel production and the high economic value of harvested biomass for biogas and fertilizer. This study found that salt concentration (∼45gL
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
92949 Trace Metal Mix A5 with Co, for microbiology, sterile