EMAIL THIS PAGE TO A FRIEND
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Alpinumisoflavone inhibits osteoclast differentiation and exerts anti-osteoporotic effect in ovariectomized mice.
PMID 28651235
Abstract
Alpinumisoflavone (AIF), a naturally occurring flavonoid compound exacted from Derris eriocarpa, has been found to have a number of pharmacological activities. However, its role in bone disorder has not been investigated. The aim of this study is to evaluate the osteoprotective effect of AIF on ovariectomy-induced bone loss in mice model and related underlying mechanisms. Our study provides experimental evidence that AIF could regulate the remodeling process of bone and exert osteoprotective effect against ovariectomy-induced bone loss. Moreover, our results show that AIF suppresses osteoclast differentiation by attenuating RANKL-induced activation of p38, ERK and JNK pathways and consequently represses the expression of c-Fos and NFATc1.
Related Materials