EMAIL THIS PAGE TO A FRIEND
Cancer discovery
Immune Escape in Breast Cancer During
PMID 28652380
Abstract
To investigate immune escape during breast tumor progression, we analyzed the composition of leukocytes in normal breast tissues, ductal carcinoma