EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
Synthesis and characterisation of robust emulsion-templated silica microcapsules.
PMID 28654882
Abstract
Robust silica microcapsules were synthesised using an emulsion template via a seeded growth strategy. Multiple additions of the silica precursor tetraethyl orthosilicate (TEOS) were observed to result in a number of physical and property changes of the capsule shells as compared to a single coating. Scanning electron microscopy indicated a morphological transition from a smooth to a roughened surface. Improved cargo retention and consolidation of the pore structure of the silica shells were observed using dye release experiments and nitrogen porosimetry respectively. In comparison to a typical hollow silica shell synthesis procedure, this one-pot loading and synthesis allows the simple production of robust capsules that are capable of sustained release, using mild conditions and reagents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
252433
Rhodamine 6G, Dye content 99 %
C28H30N2O3 · HCl