EMAIL THIS PAGE TO A FRIEND
Pharmacology, biochemistry, and behavior
Benzodiazepine-induced hyperphagia in the nondeprived rat: comparisons with CL 218,872, zopiclone, tracazolate and phenobarbital.
PMID 2865747
Abstract
Nondeprived male rats were familiarized with 30 min daily access to a highly palatable diet. Clonazepam, midazolam and chlordiazepoxide each produced significant dose-dependent increases in food consumption. Clonazepam was the most potent, and a significant hyperphagic effect was detected following 0.078 mg/kg (IP). Amongst novel non-benzodiazepine anxiolytics, zopiclone and CL 218,872 also produced significant increases in food intake. The smallest doses to produce significant hyperphagia for these two drugs were 10.0 and 2.5 mg/kg (IP) respectively. In contrast, tracazolate caused only a reduction in feeding, evident at 20 and 40 mg/kg (IP). Previous reports indicate that although benzodiazepines, zopiclone and CL 218,872 displace [3H] flunitrazepam binding in rat cerebral cortex preparations, tracazolate enhances the binding. Our results are consistent with the drug-induced hyperphagia depending upon agonist actions at high-affinity benzodiazepine sites. They also provide pharmacological evidence for a dissociation between hyperphagic and anxiolytic drug effects. Phenobarbital (2.5-40.0 mg/kg), like the benzodiazepines, produced a strong stimulation of food intake, indicating that drug action at an alternative site in the benzodiazepine receptor-GABA receptor-chloride channel complex can also lead to hyperphagia.