EMAIL THIS PAGE TO A FRIEND
The Journal of nutrition
Fish Oil Contaminated with Persistent Organic Pollutants Induces Colonic Aberrant Crypt Foci Formation and Reduces Antioxidant Enzyme Gene Expression in Rats.
PMID 28659405