EMAIL THIS PAGE TO A FRIEND
Frontiers in physiology
Human Peritoneal Mesothelial Cell Death Induced by High-Glucose Hypertonic Solution Involves Ca
PMID 28659813
Abstract
Chronic peritoneal dialysis (PD) therapy is equally efficient as hemodialysis while providing greater patient comfort and mobility. Therefore, PD is the treatment of choice for several types of renal patients. During PD, a high-glucose hyperosmotic (HGH) solution is administered into the peritoneal cavity to generate an osmotic gradient that promotes water and solutes transport from peritoneal blood to the dialysis solution. Unfortunately, PD has been associated with a loss of peritoneal viability and function through the generation of a severe inflammatory state that induces human peritoneal mesothelial cell (HPMC) death. Despite this deleterious effect, the precise molecular mechanism of HPMC death as induced by HGH solutions is far from being understood. Therefore, the aim of this study was to explore the pathways involved in HGH solution-induced HPMC death. HGH-induced HPMC death included influxes of intracellular Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5084
PTIO
C13H17N2O2