EMAIL THIS PAGE TO A FRIEND
Frontiers in plant science
The Banana Fruit SINA Ubiquitin Ligase MaSINA1 Regulates the Stability of MaICE1 to be Negatively Involved in Cold Stress Response.
PMID 28659946
Abstract
The regulation of ICE1 protein stability is important to ensure effective cold stress response, and is extensively studied in