EMAIL THIS PAGE TO A FRIEND
Reproduction in domestic animals = Zuchthygiene
Chlorogenic acid supplementation during in vitro maturation improves maturation, fertilization and developmental competence of porcine oocytes.
PMID 28660662
Abstract
Chlorogenic acid (CGA) is a quinic acid conjugate of caffeic acid, and a phytochemical found in many fruits and beverages that acts as an antioxidant. The present study investigated the effects of CGA supplementation during in vitro maturation (IVM), on in vitro development of porcine oocytes, to improve the porcine in vitro production (IVP) system. Oocytes were matured either without (control) or with CGA (10, 50, 100 and 200 μM). Subsequently, the matured oocytes were fertilized and cultured in vitro for 7 day. The rates of maturation, fertilization and blastocyst formation of oocytes matured with 50 μM CGA were significantly (p < .05) higher than those of the control oocytes. Hydrogen peroxide (H