EMAIL THIS PAGE TO A FRIEND
Applied spectroscopy
A Quantitative Fourier Transform Infrared Study of the Grafting of Aminosilane Layers on Lithium Niobate Surface.
PMID 28664784
Abstract
Due to its impressive optical properties, lithium niobate (LiNbO