EMAIL THIS PAGE TO A FRIEND
Cancer discovery
Whole-Genome and Epigenomic Landscapes of Etiologically Distinct Subtypes of Cholangiocarcinoma.
PMID 28667006
Abstract
Cholangiocarcinoma (CCA) is a hepatobiliary malignancy exhibiting high incidence in countries with endemic liver-fluke infection. We analyzed 489 CCAs from 10 countries, combining whole-genome (71 cases), targeted/exome, copy-number, gene expression, and DNA methylation information. Integrative clustering defined 4 CCA clusters-fluke-positive CCAs (clusters 1/2) are enriched in
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU025771 MISSION® esiRNA, esiRNA human RASA1 (esiRNA1)