EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Yeast Tok1p channel is a major contributor to membrane potential maintenance under chemical stress.
PMID 28669766
Abstract
Tok1p is a highly specific yeast plasma membrane potassium channel with strong outward directionality. Its opening is induced by membrane depolarization. Although the biophysical properties of Tok1p are well-described, its potentially important physiological role is currently largely unexplored. To address this issue, we examined the Tok1p activity following chemically-induced depolarization by measuring changes of plasma membrane potential (ΔΨ) using the diS-C