EMAIL THIS PAGE TO A FRIEND
Methods (San Diego, Calif.)
Radical probing of spliceosome assembly.
PMID 28669867
Abstract
Here we describe the synthesis and use of a directed hydroxyl radical probe, tethered to a pre-mRNA substrate, to map the structure of this substrate during the spliceosome assembly process. These studies indicate an early organization and proximation of conserved pre-mRNA sequences during spliceosome assembly. This methodology may be adapted to the synthesis of a wide variety of modified RNAs for use as probes of RNA structure and RNA-protein interaction.