EMAIL THIS PAGE TO A FRIEND
American journal of translational research
Valproic acid, targets papillary thyroid cancer through inhibition of c-Met signalling pathway.
PMID 28670399
Abstract
Tyrosine kinase receptors such as c-Met and its ligands are interesting therapeutic targets that have been reported to be involved in the progression of several types of cancers. Histone deacetylase inhibitor, valproic acid (VPA) is one such compound with promising anti-cancer properties. The current study was designed to evaluate the c-Met activity of VPA in thyroid carcinoma. A total 36 nu/nu mice with SW1736 cells-induced tumours were randomised into three treatment groups (5, 15, 30 mg/kg/day p.o. VPA; n = 9/group). Various cellular and enzymatic assays were performed to evaluate the dose-response relationship of VPA in c-Met inhibition.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RAB0676 Human MET / Hepatocyte Growth Factor Receptor ELISA Kit