EMAIL THIS PAGE TO A FRIEND
American journal of cancer research
Bitterness in sugar: O-GlcNAcylation aggravates pre-B acute lymphocytic leukemia through glycolysis via the PI3K/Akt/c-Myc pathway.
PMID 28670495
Abstract
Abnormal cellular energetics has emerged as a hallmark of cancer cells. Deregulating aerobic glycolysis can alter multiple metabolic and signaling pathways in cancer cells, and trigger unlimited growth and proliferation. Accumulating evidence suggests that elevated levels of protein modification with β-N-acetylglucosamine (O-GlcNAcylation) along with dysregulation of O-GlcNAc transferase (OGT) and/or O-GlcNAcase (OGA) levels may act as a nutrient sensor in cancer cells. However, the underlying mechanism of O-GlcNAcylation and the relationship between O-GlcNAcylation and glycolysis are largely unknown in pre-B acute lymphocytic leukemia (pre-B-ALL). In this study, CD19
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU082301 MISSION® esiRNA, esiRNA human OGT (esiRNA1)