EMAIL THIS PAGE TO A FRIEND
The Histochemical journal
The importance of protease histochemistry in pathology.
PMID 2867067
Abstract
The histochemical demonstration of the activities of proteases, based in the majority of cases on the use of discriminating synthetic substrates (aminoacyl or peptidyl derivatives of 4-methoxy-2-naphthylamine [MNA]), has proved very valuable in pathology. Its importance is illustrated by three topics investigated in our laboratory, namely lymphocytes, malabsorption syndrome and atherosclerosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9805
Fast Blue B Salt, Dye content ~95 %
C14H12Cl4N4O2Zn