EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Imidazole/benzimidazole-modified cyclotriphosphazenes as highly selective fluorescent probes for Cu
PMID 28671228
Abstract
Configurational isomers (cis and trans) of imidazole- or benzimidazole-modified cyclotriphosphazenes (3a, 4a or 3b, 4b) were designed, synthesized and investigated as fluorescent probes for metal ions. The newly synthesized compounds were characterized by
Related Materials