EMAIL THIS PAGE TO A FRIEND
The Journal of pathology
Clonality analysis of synchronous gastro-oesophageal junction carcinoma and distal gastric cancer by whole-exome sequencing.
PMID 28671728
Abstract
Gastro-oesophageal junction (GEJ) carcinoma and distal gastric cancer (GC) have distinct epidemiology and clinical features and their relationship is uncertain. Synchronous multiple gastric cancers located mostly at proximal and distal sites provide rare specimens for investigating the comprehensive genomic relationships among these cancers in the context of identical genetic circumstances. Formalin-fixed, paraffin-embedded (FFPE) samples from 12 patients with synchronous GEJ carcinoma and distal GC were collected in this study. Whole-exome sequencing (WES) was performed using normal tissues as a control. Mutational profiling, clonality analysis, a detailed clinico-pathological review, determination of MSI status, EBER in situ hybridization (ISH), and programmed cell death 1 (PD-1)/programmed death-ligand 1 (PD-L1) immunohistochemical staining were performed. Twenty-three of the 24 samples were microsatellite-stable (MSS). Subclonal analysis revealed that nine pairs of GEJ and distal GC tumours in neoadjuvant chemotherapy naïve patients developed independently from different origins. Two patients who received neoadjuvant chemotherapy shared clonal origins with highly similar somatic alterations. The remaining one patient who shared a rare mutation died within 6.2 months at the N3 stage. However, the enriched pathway identified from the overall mutation spectra in distal GC and GEJ carcinoma showed the close relationship of these cancers. Thus, although these cancers may have similar characteristics, histopathological and genetic profiling from single tumour specimens may still underestimate the mutational burden and somatic heterogeneity of multiple GCs. In addition, this series of cases also showed a PD-L1 expression rate of 58.3% and 66.7% in distal GC and GEJ carcinoma, respectively, with all the cases expressing PD-1. This result suggests the potential benefit of immunotherapeutic treatments. Copyright © 2017 Pathological Society of Great Britain and Ireland. Published by John Wiley & Sons, Ltd.